missing translation for 'onlineSavingsMsg'
Learn More

Carboxyphosphamide, TRC

Product Code. 30009211
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009211 5 mg 5mg
30134768 25 mg 25mg
30007940 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009211 Supplier Toronto Research Chemicals Supplier No. TRCC1813505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22788-18-7
Formula Weight 292.0146
InChI Formula InChI=1S/C7H15Cl2N2O4P/c8-2-4-11(5-3-9)16(10,14)15-6-1-7(12)13/h1-6H2,(H2,10,14)(H,12,13)
IUPAC Name 3-[amino-[bis(2-chloroethyl)amino]phosphoryl]oxypropanoic acid
Molecular Formula C7 H15 Cl2 N2 O4 P
Molecular Weight (g/mol) 293.08
SMILES NP(=O)(OCCC(=O)O)N(CCCl)CCCl
Synonym 3-[[Amino[bis(2-chloroethyl)amino]phosphinyl]oxy]propanoic Acid, N,N-Bis(2-chloroethyl)phosphorodiamidate Hydracrylic Acid, Asta 5754, Carboxycyclophosphamide, NSC 145124;
Chemical Name or Material Carboxyphosphamide

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.