Learn More
Carbutamide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30321792
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321792 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30321792 Supplier Dr. Ehrenstorfer Supplier No. DREA11041450AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Carbutamide
CAS 339-43-5
Analyte or Component Names Carbutamide
Shipping Condition Room Temperature
Molecular Formula C11 H17 N3 O3 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzenesulfonamide, 4-amino-N-[(butylamino)carbonyl]-, Sulfanilamide, N1-(butylcarbamoyl)- (6CI), Urea, 1-butyl-3-sulfanilyl- (7CI,8CI), 1-Butyl-3-sulfanilylurea, Alentin, Aminophenurobutane, BZ 55, Bucarban, Bucrol, Bukarban, Burcol, Butisulfina, Carbutamid, Carbutamide, Diaboral, Emedan, Glucidoral, Glucofren, Glybutamide, Inbuton, Invenol, N-(4-Aminobenzenesulfonyl)-N'-butylurea, N-Butyl-N'-sulfanilylurea, N-Sulfanilyl-N'-butylurea, NSC 242409, NSC 246862, Nadisan, Nadizan, Norboral, N'-(4-Aminobenzenesulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-n-butylurea, N'-(Butylcarbamoyl)sulfanilamide, Oranil, Oranyl, Orasulin, U 6987
InChI Formula InChI=1S/C11H17N3O3S/c1-2-3-8-13-11(15)14-18(16,17)10-6-4-9(12)5-7-10/h4-7H,2-3,8,12H2,1H3,(H2,13,14,15)
SMILES CCCCNC(=O)NS(=O)(=O)c1ccc(N)cc1
IUPAC Name 1-(4-aminophenyl)sulfonyl-3-butylurea
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 271.34
Quantity 1 mL
Formula Weight 271.0991 g/mol