Learn More
Carbutamide, LoGiCal
Product Code. 30462952
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462952 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462952 Supplier LoGiCal Supplier No. LGCFOR1856.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Carbutamide
CAS 339-43-5
Analyte or Component Names Carbutamide
Shipping Condition Room Temperature
Molecular Formula C11 H17 N3 O3 S
Certifications/Compliance ISO 9001
Synonym Benzenesulfonamide, 4-amino-N-[(butylamino)carbonyl]-, Sulfanilamide, N1-(butylcarbamoyl)- (6CI), Urea, 1-butyl-3-sulfanilyl- (7CI,8CI), 1-Butyl-3-sulfanilylurea, Alentin, Aminophenurobutane, BZ 55, Bucarban, Bucrol, Bukarban, Burcol, Butisulfina, Carbutamid, Carbutamide, Diaboral, Emedan, Glucidoral, Glucofren, Glybutamide, Inbuton, Invenol, N-(4-Aminobenzenesulfonyl)-N'-butylurea, N-Butyl-N'-sulfanilylurea, N-Sulfanilyl-N'-butylurea, NSC 242409, NSC 246862, Nadisan, Nadizan, Norboral, N'-(4-Aminobenzenesulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-n-butylurea, N'-(Butylcarbamoyl)sulfanilamide, Oranil, Oranyl, Orasulin, U 6987
InChI Formula InChI=1S/C11H17N3O3S/c1-2-3-8-13-11(15)14-18(16,17)10-6-4-9(12)5-7-10/h4-7H,2-3,8,12H2,1H3,(H2,13,14,15)
Solution Type Neat
SMILES CCCCNC(=O)NS(=O)(=O)c1ccc(N)cc1
IUPAC Name 1-(4-aminophenyl)sulfonyl-3-butylurea
Industry Type Clinical Testing
Molecular Weight (g/mol) 271.34
Quantity 10 mg
Formula Weight 271.0991 g/mol