Learn More
Carbutamide, Mikromol™
Description
Carbutamide
Specifications
Specifications
| Chemical Name or Material | Carbutamide |
| CAS | 339-43-5 |
| Analyte or Component Names | Carbutamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H17 N3 O3 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzenesulfonamide, 4-amino-N-[(butylamino)carbonyl]-, Sulfanilamide, N1-(butylcarbamoyl)- (6CI), Urea, 1-butyl-3-sulfanilyl- (7CI,8CI), 1-Butyl-3-sulfanilylurea, Alentin, Aminophenurobutane, BZ 55, Bucarban, Bucrol, Bukarban, Burcol, Butisulfina, Carbutamid, Carbutamide, Diaboral, Emedan, Glucidoral, Glucofren, Glybutamide, Inbuton, Invenol, N-(4-Aminobenzenesulfonyl)-N'-butylurea, N-Butyl-N'-sulfanilylurea, N-Sulfanilyl-N'-butylurea, NSC 242409, NSC 246862, Nadisan, Nadizan, Norboral, N'-(4-Aminobenzenesulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-n-butylurea, N'-(Butylcarbamoyl)sulfanilamide, Oranil, Oranyl, Orasulin, U 6987 |
| InChI Formula | InChI=1S/C11H17N3O3S/c1-2-3-8-13-11(15)14-18(16,17)10-6-4-9(12)5-7-10/h4-7H,2-3,8,12H2,1H3,(H2,13,14,15) |
| SMILES | CCCCNC(=O)NS(=O)(=O)c1ccc(N)cc1 |
| IUPAC Name | 1-(4-aminophenyl)sulfonyl-3-butylurea |
| Show More |