Learn More
Carbutamide, TRC
Product Code. 30135312
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135312 5 mg 5mg
30134490 100 mg 100mg
30040634 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135312 Supplier Toronto Research Chemicals Supplier No. TRCC1833005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 339-43-5
Molecular Formula C11 H17 N3 O3 S
Molecular Weight (g/mol) 271.34
Synonym Benzenesulfonamide, 4-amino-N-[(butylamino)carbonyl]-, Sulfanilamide, N1-(butylcarbamoyl)- (6CI), Urea, 1-butyl-3-sulfanilyl- (7CI,8CI), 1-Butyl-3-sulfanilylurea, Alentin, Aminophenurobutane, BZ 55, Bucarban, Bucrol, Bukarban, Burcol, Butisulfina, Carbutamid, Carbutamide, Diaboral, Emedan, Glucidoral, Glucofren, Glybutamide, Inbuton, Invenol, N-(4-Aminobenzenesulfonyl)-N'-butylurea, N-Butyl-N'-sulfanilylurea, N-Sulfanilyl-N'-butylurea, NSC 242409, NSC 246862, Nadisan, Nadizan, Norboral, N'-(4-Aminobenzenesulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-butylurea, N'-(4-Aminophenylsulfonyl)-N-n-butylurea, N'-(Butylcarbamoyl)sulfanilamide, Oranil, Oranyl, Orasulin, U 6987
IUPAC Name 1-(4-aminophenyl)sulfonyl-3-butylurea
SMILES CCCCNC(=O)NS(=O)(=O)c1ccc(N)cc1
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C11H17N3O3S/c1-2-3-8-13-11(15)14-18(16,17)10-6-4-9(12)5-7-10/h4-7H,2-3,8,12H2,1H3,(H2,13,14,15)
Formula Weight 271.0991
Percent Purity >95%
Chemical Name or Material Carbutamide
Grade HPLC