missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carcinoembryonic Antigen / CD66 Mouse, Clone: C66/195, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1048-MSM2-P1
This item is not returnable.
View return policy
Specifications
| Carcinoembryonic Antigen / CD66 | |
| Cytoplasmic, luminal surface | |
| C66/195 | |
| Unconjugated | |
| NP_011525508 | |
| Mouse | |
| 80-200kDa | |
| Protein A/G Purified | |
| RUO | |
| 634 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| CEACAM1 and CEACAM5 | |
| Recombinant full-length human CEA protein | |
| MCF7 or 293T cells. Colon carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction