missing translation for 'onlineSavingsMsg'
Learn More

Carcinoembryonic Antigen / CD66 Mouse, Clone: COL-1, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 1048-MSM1-P1

Product Code. 15909737

  • ONLINE EXCLUSIVE PRICE
    € 554.40
    € 478.00 / 100µg
    Save € 76.40
    13% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Carcinoembryonic Antigen / CD66
Cytoplasmic, luminal surface
COL-1
Unconjugated
NP_011525508
Mouse
80-200kDa
Protein A/G Purified
RUO
634
IgG2a κ
Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide
CEACAM1 and CEACAM5
Human colon carcinoma extract
MCF7 or 293T cells. Colon carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.