missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD10 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76676DL594
This item is not returnable.
View return policy
Description
CARD10 Polyclonal antibody specifically detects CARD10 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CARD10 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 594 | |
| CARD-containing MAGUK 3 protein, CARD-containing MAGUK protein 3, Carma 3, CARMA3BIMP1Bcl10 binding protein and activator of NFKB, caspase recruitment domain family, member 10, caspase recruitment domain-containing protein 10, MGC142219 | |
| Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the C-terminus of human CARD10. The immunogen is located within the last 50 amino acids of CARD10. Amino Acid Squence: KECGSSRGCPSSSED | |
| 0.1 mL | |
| Apoptosis | |
| 29775 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction