missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARD12 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-225AF594
This item is not returnable.
View return policy
Description
CARD12 Polyclonal antibody specifically detects CARD12 in Human samples. It is validated for Western Blot
Specifications
| CARD12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| CARD12leucine rich repeat and CARD domaincontaining 4, caspase recruitment domain family, member 12, Caspase recruitment domain-containing protein 12, Clan protein, CLAN1ipaf, CLANCARD, LRR, and NACHT-containing protein, CLR2.1, Ice protease-activating factor, Ipaf, NLR family, CARD domain containing 4 | |
| N terminus (MNFIKDNSRALIQRMGM) of the A and B isoforms of the human CLAN protein. | |
| 0.1 mL | |
| Apoptosis, Immunology, Innate Immunity | |
| 58484 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction