missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CARD6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13402637
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13402637 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13402637 Supplier Invitrogen™ Supplier No. PA528517

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARD6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene CARD6
Gene Accession No. Q9BX69
Gene Alias Card6; CARD-containing inhibitor of Nod1 and Cardiak-induced NF-kB activation; caspase recruitment domain family member 6; caspase recruitment domain family, member 6; caspase recruitment domain protein 6; caspase recruitment domain-containing protein 6; CINCIN1; D730008L15
Gene Symbols CARD6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human CARD6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84674
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.