missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122163
This item is not returnable.
View return policy
Description
CARD9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined.
Specifications
| CARD9 | |
| Polyclonal | |
| Unconjugated | |
| CARD9 | |
| CANDF2; card 9; Card9; caspase recruitment domain family member 9; caspase recruitment domain family, member 9; caspase recruitment domain protein 9; caspase recruitment domain-containing protein 9; Gm782; hCARD9; rCARD9; RP11-413M3.9 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 64170 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9H257 | |
| CARD9 | |
| Recombinant protein Caspase Recruitment Domain Family, Member 9. The antigen corresponds to amino acid range 26-283 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction