Learn More
Tenak™ Cardboard Cryoboxes
Shop All TENAK Products
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Dimensions (L x W x H):
75 x 75 mm
100 x 100 mm
133 x 133 x 32 mm
133 x 133 x 50 mm
133 x 133 x 52 mm
133 x 133 x 75 mm
136 x 136 x 32 mm
136 x 136 x 50 mm
136 x 136 x 75 mm
133 x 133 x 100 mm
133 x 133 x 130 mm
Includes:
10 x 10 Divider
9 x 9 Divider
Dividers
Unit Size:
Pack of 36
Description
- Cardboard
- Temperature resistant down to -80°C
- Ideal for ultra-low temperature storage
Specifications
Specifications
| Color | Red |
| Material | Cardboard |
| Shape | Square |
| Type | Cryo Box |
| For Use With (Application) | Cryogenic Storage of Liquids |
| Includes | Dividers |
| Quantity | 36 Pack |
| Dimensions (L x W x H) | 133 x 133 x 32 mm |
| Length (Metric) | 133 mm |
| Width (Metric) | 133 mm |
| Show More |