missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Rat anti-Human, Mouse, Rat, Drosophila, Fungi, Yeast, Biotin, Clone: YOL1/34, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-1639B-0.25ML
This item is not returnable.
View return policy
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Drosophila, Fungi, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
| alpha Tubulin | |
| Monoclonal | |
| Biotin | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| This alpha Tubulin Antibody (YOL1/34) was made from yeast tubulin. | |
| 0.25 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| 7846 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| YOL1/34 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Drosophila, Fungi, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction