missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cardiac Troponin T Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102358
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TNNT2.
Cardiac Troponin T is the tropomyosin-binding subunit of the troponin complex, which is located on the thin filament of striated muscles and regulates muscle contraction in response to alterations in intracellular calcium ion concentration. Cardiac Troponin T is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. Cardiac Troponin T expression in the human heart is affected by heart failure. Mutations within the Cardiac Troponin T protein have been associated with familial hypertrophic cardiomyopathy as well as with dilated cardiomyopathy. Transcripts for the Cardiac Troponin T gene undergo alternative splicing that results in many tissue-specific isoforms.
Specifications
| Cardiac Troponin T | |
| Polyclonal | |
| Unconjugated | |
| TNNT2 | |
| alternatively spliced; cardiac muscle troponin T; cardiac TnT; cardiac troponin 2; cardiac troponin 2 adult #1 isoform; cardiac troponin 2 adult #2 isoform; cardiac troponin 2 embryonic #1 isoform; cardiac troponin 2 embryonic #2 isoform; cardiac troponin 2, exclusion exon 6 #1 isoform; cardiac troponin 2, exclusion exon 6 #2 isoform; cardiac troponin T; cardiac troponin T form I; cardiac troponin T isoform 1; cardiac troponin T isoform 2; cardiac troponin T isoform I; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, hypertrophic 2; CMD1D; CMH2; CMPD2; cTnT; Ctt; CTTG; LVNC6; MGC3889; RATCTTG; RCM3; TNNT2; Tnnt3; Tnt; TnTc; troponin T; Troponin T cardiac; troponin T cardiac isoform; troponin T isoform 1; troponin T isoform 2; troponin T splicing mutant isoform 1; troponin T splicing mutant isoform 2; troponin T type 2 (cardiac); troponin T, cardiac muscle; Troponin T, cardiac muscle isoforms; Troponin T2; troponin T2, cardiac; troponin T2, cardiac type; truncated cardiac troponin T | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21956, 24837, 7139 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P45379, P50752, P50753 | |
| TNNT2 | |
| A synthesized peptide derived from human TNNT2(Accession P45379), corresponding to amino acid residues A255-K298. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction