missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CARF Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148815
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
CARF (calcium-response factor), also known as ALS2CR8 (amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein) or NYD-SP24, is a 725 amino acid nuclear protein. Expressed in a wide variety of tissues with highest expression in the hippocampus, CARF is thought to be a transcription factor that associates with the p53 tumor suppression pathway. CARF cooperates, co-localizes and is co-regulated with ARF, an ADP-ribosylation factor, and, through this interaction, helps to mediate ARF-p53-induced apoptotic signaling. This apoptotic pathway is implicated in cell cycle control, proper cellular development, response to DNA damage and the aging process, suggesting that CARF participates in various events throughout the cell.
Specifications
| CARF | |
| Polyclonal | |
| Unconjugated | |
| CARF | |
| Als2cr8; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 8; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 8 protein homolog; calcium response factor; calcium responsive transcription factor; calcium-response factor; calcium-response factor CaRF; calcium-responsive transcription factor; CaRF; ECBRC-FC1; ECBRC-FC2; NYD-SP24; testis development protein NYD-SP24 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 79800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8N187 | |
| CARF | |
| E. coli-derived recombinant human CARF, Ser379-Leu579. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction