missing translation for 'onlineSavingsMsg'
Learn More

Carglumic Acid, MedChemExpress

Product Code. 16471436
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471436 200 mg 200mg
16492196 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471436 Supplier Medchem Express Supplier No. HYB0711

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carglumic acid (N-Carbamyl-L-glutamic acid), a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.

Carglumic acid (N-Carbamyl-L-glutamic acid), a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.

Specifications

Chemical Name or Material Carglumic Acid
Physical Form Solid
CAS 1188-38-1
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C6H10N2O5
For Use With (Application) Cancer-programmed cell death
Synonym N-Carbamyl-L-glutamic acid
Solubility Information DMSO : 100 mg/mL (525.90 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (43.81 mM; Need ultrasonic)
SMILES O=C(O)CC[C@@H](C(O)=O)NC(N)=O
Molecular Weight (g/mol) 190.15
Formula Weight 190.15
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.