missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carglumic Acid, MedChemExpress
Description
Carglumic acid (N-Carbamyl-L-glutamic acid), a functional analogue of N-acetylglutamate (NAG) and a carbamoyl phosphate synthetase 1 (CPS1) activator, is used to treat acute and chronic hyperammonemia associated with NAG synthase (NAGS) deficiency.
Specifications
Specifications
| Chemical Name or Material | Carglumic Acid |
| Physical Form | Solid |
| CAS | 1188-38-1 |
| Color | White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C6H10N2O5 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | N-Carbamyl-L-glutamic acid |
| Solubility Information | DMSO : 100 mg/mL (525.90 mM; Need ultrasonic) ∣H2O : 8.33 mg/mL (43.81 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?