missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARHSP1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22260C
This item is not returnable.
View return policy
Description
CARHSP1 Polyclonal antibody specifically detects CARHSP1 in Human samples. It is validated for Western Blot
Specifications
| CARHSP1 | |
| Polyclonal | |
| Western Blot | |
| calcium regulated heat stable protein 1 (24kD), calcium regulated heat stable protein 1, 24kDa, calcium-regulated heat stable protein 1, calcium-regulated heat-stable protein (24kD), Calcium-regulated heat-stable protein of 24 kDa, CRHSP-24MGC111446, CSDC1 | |
| The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Calcium Regulated Heat Stable Protein 1 using the numbering given in entry NP_055131.2 (GeneID 23589). | |
| 0.1 mL | |
| Signal Transduction | |
| 23589 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction