missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARKL Antibody (OTI3D7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71771V
This item is not returnable.
View return policy
Description
CARKL Monoclonal specifically detects CARKL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CARKL | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SHPK | |
| Full length recombinant protein of human CARKL (NP_037408) produced in HEK293T cells. | |
| 0.1 mL | |
| 23729.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3D7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| carbohydrate kinase-like, Carbohydrate kinase-like protein, CARKLEC 2.7.1.14, sedoheptulokinase, SHKFLJ32478 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction