Learn More
CARKL Antibody (OTI3D7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71771G
Product Code. 18109654
  • € 751.00 / 0.10mL
Description
CARKL Monoclonal specifically detects CARKL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CARKL
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
SHPK
Full length recombinant protein of human CARKL (NP_037408) produced in HEK293T cells.
0.1 mL
23729.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI3D7
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
carbohydrate kinase-like, Carbohydrate kinase-like protein, CARKLEC 2.7.1.14, sedoheptulokinase, SHKFLJ32478
Mouse
Affinity Purified
Primary
Human, Mouse, Rat