Learn More
Carl Zeiss™ C-mount Camera Adapter
Product Code. 11826442
Click to view available options
For Use With (Equipment):
C mount camera
Unit Size:
Each
Product Code. 11826442
Brand: Carl Zeiss™ 0000001069414
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Adapts specific Zeiss microscopes for use with still or video camera
Adapts microscope for use with C-mount still or video cameras; 0.63X f/60mm interface
Accessory for:
  • Stemi 2000 C /CS
  • Axiostar
  • Axioskop
  • AxioImager D1
Specifications
Type Camera Adapter
For Use With (Equipment) C mount camera
Required Accessories Stemi™ 2000C/CS, Axiostar™, Axioskop™, AxioImager D1