Learn More
Carl Zeiss™ Neutral-Density Filter
Suitable for a range of Axio microscopes
Brand:  Carl Zeiss™ 467848-0000-000
Additional Details : Weight : 0.00450kg
Product Code. 10649138
  • € 26.00 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
N = 0.06; D = 32 x 2
Compatible with the following microscopes:
  • Axio Imager (A1, A1m, D1, D1 for epi-fluorescence with ApoTome equipment, D1m, M1, M1m, Z1, Z1 + ApoTome, Z1m)
  • AxioLab™ (A1, A1 FL-LED, A1 MAT, A1 Pol, A1 Entry)
  • Axio Observer (D1, Observer.D1 Entry, D1 Mid Range, Z1, Z1 Cell Observer, Z1 High End, Z1m ACR)
  • Axio Scope 1 Pol
  • AxioLab
  • Axioplan 2 Imaging (Imaging E, Imaging for epi-fluorescence with ApoTome equipment, Imaging MAT)
  • Axioskop™ 2 (FS MOT, FS Plus, MAT, MAT mot, MOT Plus, Plus)
  • Axioskop (20, 40, 40A Pol, 40FL, 40 Pol, Axioskop 50)
  • Axiostar™ (plus, plus FL, vario)
  • Axiovert™ (25, 25C, 25CFL, 40C, 40CFL)
  • PALM CombiSystem Rel. 4.2 and MicroBeam Rel.4.2
  • LM MicroTweezers Rel.4.2
Specifications
Specifications
Axio Imager, AxioLab™, Axio Observer, Axio Scope 1 Pol, AxioLab™, Axioplan 2 Imaging, Axioskop™ 2, Axioskop™, Axiostar™, Axiovert™ microscopes, PAL