Learn More
Motic™ CCIS™ Plan Achromatic Phase Objective
Provides a flat field of view while also correcting for color aberration. Motic™ CCIS™ Plan Achromatic Phase Objective is useful on specimens that do not absorb light.
Brand: Motic™ 1101001703281
Additional Details : Weight : 0.01000kg
Description
- For use with AE31 Inverted Microscope
- Achromatic Plan Phase Objective
- Flat field
- Eliminates color aberration
- Magnification and Numerical Apertures available:
- 10X/0.25 (WD=4.1mm) Ph1 for AE
- 4X/0.10 (WD=12.6mm) Ph0 for AE
Specifications
PL Ph0 4X | |
0.1 mm, WD=12.6 mm | |
AE31 inverted microscope | |
1 Piece |