missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carm1 Antibody (3H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37645-0.025ml
This item is not returnable.
View return policy
Description
Carm1 Monoclonal specifically detects Carm1 in Human, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Carm1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| CARM1 | |
| Purified recombinant fragment of human CARM1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Rat, Monkey, Primate | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3H2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
| Mouse | |
| 65 kDa | |
| 0.025 mL | |
| Cancer, Cellular Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 10498 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction