missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carm1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98754CL1
This item is not returnable.
View return policy
Description
Carm1 Polyclonal antibody specifically detects Carm1 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Carm1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Carm1. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 10498 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction