missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carm1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-341
This item is not returnable.
View return policy
Description
Carm1 Polyclonal specifically detects Carm1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Carm1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB200-341 is specific for human CARM1 protein. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:5000-1:15000, Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 1-4 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q86X55 | |
| CARM1 | |
| A synthetic peptide that maps to a region between residues 450 and 500 of human Coactivator-Associated Arginine Methyltransferase 1 using the numbering given in Swiss-Prot entry Q86X55 (GeneID 10498). | |
| 100 μL | |
| Cancer, Cellular Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 10498 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction