missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Carm1 Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB200-341PE
This item is not returnable.
View return policy
Description
Carm1 Polyclonal antibody specifically detects Carm1 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Carm1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| PE | |
| coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4 | |
| A synthetic peptide that maps to a region between residues 450 and 500 of human Coactivator-Associated Arginine Methyltransferase 1 using the numbering given in Swiss-Prot entry Q86X55 (GeneID 10498). | |
| 0.1 mL | |
| Cancer, Cellular Markers, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 10498 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction