Learn More
STEMCELL Technologies Carmine Alum
Product Code. 17189231
Encompass_Preferred
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 17189231
Brand: STEMCELL Technologies 07070
Dye for staining mouse mammary gland whole mounts
  • Carmine Alum is used for the whole mount staining of mouse mammary gland
  • This product contains carmine and aluminum potassium sulfate in water
Specifications
Type Carmine Alum
For Use With (Application) Immunohistochemistry
Quantity 100 mL