missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CARNMT1 Antibody (OTI4B7), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72398PECY55
This item is not returnable.
View return policy
Description
CARNMT1 Monoclonal specifically detects CARNMT1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CARNMT1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| CARNMT1 | |
| Full length recombinant protein of human C9orf41 (NP_689663) produced in HEK293T cells. | |
| 0.1 mL | |
| 138199.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4B7 | |
| Flow Cytometry | |
| C9orf41, chromosome 9 open reading frame 41, FLJ25795, hypothetical protein LOC138199 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction