Learn More
CARS Antibody (8B11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26466-50ul
 View more versions of this product
Product Code. 30031154
  • € 343.00 / 50µL
Description
CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CARS
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
8B11
Western Blot 1:500-1:2000, ELISA
CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246
A synthesized peptide derived from Human CARS [UniProt P49589]
50 μL
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
833
Store at -20 to -70C. Avoid freeze-thaw cycles.