Learn More
Invitrogen™ CARS Recombinant Rabbit Monoclonal Antibody (24GB3015)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557010
 View more versions of this product
Product Code. 30275918
  • € 146.00 / 20µL
Description
CARS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CARS
Recombinant Monoclonal
Unconjugated
CARS1
CA3; CARS; cars protein; CARS1; CYSRS; cysteine translase; cysteine tRNA ligase 1, cytoplasmic; cysteine--tRNA ligase, cytoplasmic; Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase 1; cysteinyl-tRNA synthetase, cytoplasmic; LOW QUALITY PROTEIN: cysteine--tRNA ligase, cytoplasmic; MGC:11246; sb:cb71
Rabbit
Affinity chromatography
RUO
293638, 833
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3015
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49589
CARS, CARS1
A synthesized peptide derived from human CARS (aa 650-748).
20 μL
Primary
Human, Rat