missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol, MedChemExpress

Product Code. 16421996
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421996 500 mg 500mg
16458056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16421996 Supplier Medchem Express Supplier No. HYB0006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carvedilol (BM 14190) is a non-selective β/α-1 blocker. Carvedilol inhibits lipid peroxidation in a dose-dependent manner with an IC50 of 5 μM. Carvedilol is a multiple action antihypertensive agent with potential use in angina and congestive heart failure. Carvedilol is an autophagy inducer that inhibits the NLRP3 inflammasome.

Carvedilol (BM 14190) is a non-selective β/α-1 blocker. Carvedilol inhibits lipid peroxidation in a dose-dependent manner with an IC50 of 5 μM. Carvedilol is a multiple action antihypertensive agent with potential use in angina and congestive heart failure. Carvedilol is an autophagy inducer that inhibits the NLRP3 inflammasome.

Specifications

Chemical Name or Material Carvedilol
CAS 72956-09-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H26N2O4
Quantity 500 mg
Synonym BM 14190
Solubility Information DMSO : 100 mg/mL (246.02 mM; Need ultrasonic)
SMILES OC(CNCCOC1=CC=CC=C1OC)COC2=CC=CC(N3)=C2C4=C3C=CC=C4
Molecular Weight (g/mol) 406.47
Formula Weight 406.47
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.