Learn More
Cas9 Monoclonal Antibody (OTI1C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811327S
Encompass_Preferred
 View more versions of this product
Product Code. 17575244
  • € 242.00 / 30µL
Description
Cas9 Monoclonal antibody specifically detects Cas9 in Human samples. It is validated for Immunoprecipitation
Specifications
Specifications
Cas9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8WZ75
Mouse
Affinity chromatography
RUO
54538
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunoprecipitation
OTI1C8
Unconjugated
ROBO4
cas9; CASP 9; Caspase9; Caspase-9 precursor; CASPASE-9c; CRISPR9; CRISPR-Cas9; CTD; OTTHUMP00000002324; RNCASP9; RP11-265F14.3; s aureus
Human recombinant protein fragment corresponding to amino acids 32-331 of human ROBO4 produced in E.coli.