missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cas9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143568
This item is not returnable.
View return policy
Description
Cas9 Polyclonal Antibody for Western Blot, ICC/IF
Cas9 is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria and genetic engineering. The system is unique and flexible due to its dependence on RNA as the element that targets the nuclease to a desired DNA sequence. It can be used to induce indel mutations, specific sequence replacements/insertions, and large deletions or genomic rearrangements at any desired location in the genome. In addition, Cas9 can also be used to mediate upregulation of specific endogenous genes or to alter histone modifications or DNA methylation. Cas9 may be derived from Streptococcus pyogenes (SpCas9) or Staphylococcus aureus (SaCas9). SaCas9 has a number of properties that make it advantageous for genome editing, including its small size, high efficiency, nickase activity, and apparent specificity. SaCas9 principally recognizes an NNGRRT protospacer adjacent motif (PAM) and cleaves target DNA at rates comparable to SpCas9 with an ideal spacer lengths of 24 to 20 nucleotides. In comparison to SpCas9, SaCas9 is smaller (1368 AA versus 1053 AA) and generates indels at a comparable rate when directed against target DNA with the mutually permissive NGGRRT PAM. This makes it the preferred enzyme for all-in-one delivery of Cas9 and multiple gRNA expression cassettes with Adeno-associated vectors.
Specifications
| Cas9 | |
| Polyclonal | |
| Unconjugated | |
| cas9; CRISPR9; CRISPR-Cas9; s aureus | |
| CAS9 from S. aureus expressed in and purified from E. coli. | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot | |
| Conc. Not Determined | |
| whole serum with no preservative | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction