missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Casamino Acids

Supplemented to microbiological growth media.

Brand:  MP Biomedicals™ 113060012

Additional Details : CAS : 91079-40-2 Weight : 0.22700kg

 View more versions of this product

Product Code. 11307719

  • € 350.86 / 227g
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Applications: Casamino acids is supplemented to microbiological growth media.
Product Description: Casamino acids are a mixture of amino acids (except tryptophan) obtained from acid hydrolysis of casein.
Specifications

Specifications

Store at Room Temperature (15-30 °C).
Powder
Media Supplement
227 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers