missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CASD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569624
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
O-acetyltransferase that catalyzes 9-O-acetylation of sialic acids (PubMed:20947662, PubMed:26169044). Sialic acids are sugars at the reducing end of glycoproteins and glycolipids, and are involved in various processes such as cell-cell interactions, host-pathogen recognition (PubMed:20947662, PubMed:26169044).
Specifications
| CASD1 | |
| Polyclonal | |
| Unconjugated | |
| CASD1 | |
| C7orf12; Cas1; CAS1 domain containing 1; CAS1 domain-containing protein 1; Casd1; Cast1; N-acetylneuraminate 9-O-acetyltransferase; Nbla04196; O-acetyltransferase; Sialate O-acetyltransferase; SOAT; WUGSC:H_GS542D18.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 64921 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96PB1 | |
| CASD1 | |
| synthetic peptide directed towards the N terminal region of human CASD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction