missing translation for 'onlineSavingsMsg'
Learn More

Casein, High Nitrogen

Product Code. 15885098 Shop All Thermo Scientific Products
Click to view available options
Quantity:
11.3 kg (25 lb.)
2.2 kg (5 lb.)
Unit Size:
11339.8g
2268g
This item is not returnable. View return policy

Product Code. 15885098

Brand: Thermo Scientific J12845.=A

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prepared from New Zealand casein exclusively.

  • Format: White to off-white powder
  • Protein (Dry basis): ≥95.0%
  • Melting point: ∽280°C (dec.)
  • MDLNumber: MFCD00081481
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY

Specifications

Name Casein
Quantity 2.2 kg (5 lb.)
Packaging Plastic bottle
Sensitivity Hygroscopic
Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.