missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1, Gamma 2 Rabbit Anti-Human,Mouse, Polyclonal, Abnova&Trade;
Rabbit Polyclonal Antibody
Brand: Abnova PAB2879.400uL
This item is not returnable.
View return policy
Description
casein kinase 1, gamma 2 Polyclonal antibody specifically detects casein kinase 1, gamma 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| casein kinase 1, gamma 2 | |
| Polyclonal | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. |
|
| CK1g2 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CSNK1G2. | |
| RUO | |
| 1455 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| In PBS (0.09% sodium azide) | |
| Rabbit | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction