missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 1 gamma Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32245DL594
This item is not returnable.
View return policy
Description
Casein Kinase 1 gamma Polyclonal antibody specifically detects Casein Kinase 1 gamma in Human, Mouse samples. It is validated for Western Blot
Specifications
| Casein Kinase 1 gamma | |
| Polyclonal | |
| Western Blot | |
| casein kinase 1, gamma 1, casein kinase I isoform gamma-1, CKI-gamma 1, EC 2.7.11, EC 2.7.11.1 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Casein Kinase I Isoform gamma-1 using the numbering given in entry NP_071331.2 (GeneID 53944). | |
| 0.1 mL | |
| Protein Kinase | |
| 53944 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction