Learn More
Casein Kinase 2 alpha Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-378FR
Product Code. 18750000
  • € 633.00 / 0.10mL
Description
Casein Kinase 2 alpha Polyclonal antibody specifically detects Casein Kinase 2 alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry (Paraffin)
DyLight 680
The immunogen recognized by this antibody maps to a region between residue 350 and the C-terminus (residue 391) of human Casein Kinase 2, alpha 1 using the numbering given in entry NP_001886.1 (GeneID 1457).
1457
IgG
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin