missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Casein Kinase 2 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 302.00 - € 498.00
Specifications
| Antigen | Casein Kinase 2 alpha |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289393
|
Novus Biologicals
NBP2-57124 |
100 μL |
€ 498.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661017
|
Novus Biologicals
NBP2-57124-25ul |
25 μL |
€ 302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Casein Kinase 2 alpha Polyclonal specifically detects Casein Kinase 2 alpha in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Casein Kinase 2 alpha | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, DNA Repair, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 1457 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SGISSVPTPSPLGPLAGSPVIAAANPLGMPVPAAAG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| casein kinase 2, alpha 1 polypeptide, casein kinase II alpha 1 subunit, CK II alpha, CK2 catalytic subunit alpha, CK2A1casein kinase II subunit alpha, CKII, EC 2.7.11, EC 2.7.11.1, protein kinase CK2 | |
| CSNK2A1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title