missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASK Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-41181
This item is not returnable.
View return policy
Description
CASK Polyclonal specifically detects CASK in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CASK | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CASK | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CASK. The immunogen is located within amino acids 760 - 810 of CASK. Amino Acid Squence: YPIPHTTRPPKKDEENGKN | |
| 0.1 mg | |
| Signal Transduction | |
| 8573 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 ug/mL, Simple Western, ELISA, Immunocytochemistry/Immunofluorescence 20 ug/mL | |
| Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least six alternatively spliced transcript variants have been observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction