missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASK Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41181MFV500
This item is not returnable.
View return policy
Description
CASK Polyclonal antibody specifically detects CASK in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| CASK | |
| Polyclonal | |
| Western Blot, Simple Western, ELISA, Immunocytochemistry/ Immunofluorescence | |
| Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human CASK. The immunogen is located within amino acids 760 - 810 of CASK. Amino Acid Squence: YPIPHTTRPPKKDEENGKN | |
| 0.1 mL | |
| Signal Transduction | |
| 8573 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction