missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CASKIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563228
This item is not returnable.
View return policy
Description
Immunogen sequence: GKEQELVQAV KAEDVGTAQR LLQRPRPGKA KLLGSTKKIN VNFQDPDGFS A Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Calcium/calmodulin-dependent serine protein kinase (CASK) is a conserved multi-domain scaffolding protein involved in brain development, synapse formation, and establishment of cell polarity. CASKINs (CASK interacting proteins) interact with CASK and is thought to play a role in CASK function, specifically by coupling CASK to distinct downstream effectors. CASKIN1 is a multidomain protein containing six N-terminal ankyrin repeats, one SH3 domain, and two sterile alpha motif domains followed by a long proline-rich sequence and a short conserved C-terminal domain. CASKIN1 coassembles with CASK on the cytoplasmic tail of neurexin 1, suggesting that CASKIN and CASK coat the cytoplasmic tails of neurexins and other cell-surface proteins.
Specifications
| CASKIN1 | |
| Polyclonal | |
| Unconjugated | |
| CASKIN1 | |
| ANKS5A; CASK interacting protein 1; CASKIN1; Caskin-1; CASK-interacting protein 1; KIAA1306 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WXD9 | |
| CASKIN1 | |
| Recombinant protein corresponding to Human CASKIN1. Recombinant protein control fragment (Product #RP-101673). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction