missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CASKIN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534424
This item is not returnable.
View return policy
Description
A suggested positive control is Hela cell lysate. PA5-34424 can be used with blocking peptide PEP-1466.
Calcium/calmodulin-dependent serine protein kinase (CASK) is a conserved multi-domain scaffolding protein involved in brain development, synapse formation, and establishment of cell polarity. CASKINs (CASK interacting proteins) interact with CASK and is thought to play a role in CASK function, specifically by coupling CASK to distinct downstream effectors. CASKIN1 is a multidomain protein containing six N-terminal ankyrin repeats, one SH3 domain, and two sterile alpha motif domains followed by a long proline-rich sequence and a short conserved C-terminal domain. CASKIN1 coassembles with CASK on the cytoplasmic tail of neurexin 1, suggesting that CASKIN and CASK coat the cytoplasmic tails of neurexins and other cell-surface proteins.
Specifications
| CASKIN1 | |
| Polyclonal | |
| Unconjugated | |
| CASKIN1 | |
| ANKS5A; CASK interacting protein 1; CASKIN1; Caskin-1; CASK-interacting protein 1; KIAA1306 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57524 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8WXD9 | |
| CASKIN1 | |
| A 17 amino acid synthetic peptide near the center of human CASKIN1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction