missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASKIN2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52629AF594
This item is not returnable.
View return policy
Description
CASKIN2 Polyclonal antibody specifically detects CASKIN2 in Human samples. It is validated for Western Blot
Specifications
| CASKIN2 | |
| Polyclonal | |
| Western Blot | |
| ANKS5B, CASK interacting protein 2, caskin-2, FLJ21609, KIAA1139 | |
| The immunogen for this product maps to a region between residue 1100 to 1150 of human CASK Interacting Protein 2 using the numbering given in entry NP_065804.2 (GeneID 57513). | |
| 0.1 mL | |
| Signal Transduction | |
| 57513 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction