missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASKIN2 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812217S
This item is not returnable.
View return policy
Description
CASKIN2 Monoclonal antibody specifically detects CASKIN2 in Human samples. It is validated for Western BlotSpecifications
| CASKIN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q92614 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 399687 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI4A3 | |
| Unconjugated | |
| MYO18A | |
| 1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139 | |
| Human recombinant protein fragment corresponding to amino acids 600-1000 of human MYO18A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction