missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASKIN2 Monoclonal Antibody (OTI4A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812217
This item is not returnable.
View return policy
Description
CASKIN2 encodes a large protein that contains six ankyrin repeats, as well as a Src homology 3 (SH3) domain and two sterile alpha motif (SAM) domains, which may be involved in protein-protein interactions. The C-terminal portion of this protein is proline-rich and contains a conserved region. A related protein interacts with calcium/calmodulin-dependent serine protein kinase (CASK). Alternative splicing results in multiple transcript variants.Specifications
| CASKIN2 | |
| Monoclonal | |
| Unconjugated | |
| CASKIN2 | |
| ANKS5B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57513 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4A3 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WXE0 | |
| CASKIN2 | |
| Human recombit protein fragment corresponding to amino acids 147-552 of human CASKIN2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction