missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-2 Antibody (10C6), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80095C
This item is not returnable.
View return policy
Description
Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Specifications
| Caspase-2 | |
| Monoclonal | |
| DyLight 650 | |
| caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57 | |
| Recombinant human caspase-2 (p19 subunit). | |
| 0.1 mL | |
| Apoptosis, Cancer, Caspases | |
| 835 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| 10C6 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction