missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-2 Antibody (3D4), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Caspase-2 |
|---|---|
| Clone | 3D4 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025594
|
Novus Biologicals
NBP3-26465-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30027331
|
Novus Biologicals
NBP3-26465-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caspase-2 Monoclonal antibody specifically detects Caspase-2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Caspase-2 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57 | |
| A synthesized peptide derived from Human Caspase-2 [UniProt P42575] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3D4 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Caspases | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 835 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title